siRNA Details
Gene Symbol: Fam212b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163356.1 | NP_001156828.1 | 2 | 634 | ||
NM_175398.4 | NP_780607.2 | 2 | 781 | |||
GenBank mRNA | AK033558.1 | - | 2 | 714 | ||
AK078368.1 | - | 2 | 634 | |||
BC052371.1 | - | 2 | 714 | 6491043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
