siRNA Details
Gene Symbol: Pfdn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013369.2 | NP_001013387.1 | 2 | 260 | ||
NM_001110152.2 | NP_001103622.1 | 2 | 328 | |||
NM_001199902.1 | NP_001186831.1 | 2 | 328 | |||
GenBank mRNA | AK007940.1 | - | 2 | 122 | ||
AK136249.1 | - | 2 | 329 | |||
BC048510.1 | - | 2 | 112 | |||
BC065407.1 | - | 2 | 260 | |||
BC147673.1 | - | 2 | 161 | 9056646 | ||
BC147674.1 | - | 2 | 161 | 9056647 | ||
BU503885.1 | - | 1 | 99 |

More Information
Other Species Targeted

Gene Ontology Categories:
