siRNA Details
Gene Symbol: Ints5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176843.3 | NP_789813.1 | 2 | 2264 | ||
GenBank mRNA | AK075663.1 | - | 1 | 613 | ||
AK143868.1 | - | 2 | 2227 | |||
AK144770.1 | - | 2 | 2239 | |||
AK155499.1 | - | 2 | 2226 | |||
AK166149.1 | - | 2 | 2237 | |||
AK168795.1 | - | 2 | 2237 | |||
AK170584.1 | - | 2 | 2226 | |||
AK183009.1 | - | 1 | 396 | |||
BC039214.1 | - | 2 | 2246 | 4191047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
