siRNA Details
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175414.4 | NP_780623.1 | 7, 8 | 749 | ||
GenBank mRNA | AK079139.1 | - | 7, 8 | 722 | ||
AK158729.1 | - | 6, 7 | 634 | |||
AK163395.1 | - | 7, 8 | 805 | |||
AK203288.1 | - | 1, 2 | 95 | |||
BC052503.1 | - | 7, 8 | 731 | 5146854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
