siRNA Details
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146061.4 | NP_666173.4 | 4 | 487 | ||
GenBank mRNA | AF482997.1 | - | 4 | 482 | ||
AK046057.1 | - | 5 | 304 | |||
AK081231.1 | - | 4 | 483 | |||
BC024991.1 | - | 5 | 371 | 5065129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
