siRNA Details
Gene Symbol: C1qtnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135172.1 | NP_001128644.1 | 3 | 1074 | ||
NM_175425.4 | NP_780634.2 | 3 | 717 | |||
GenBank mRNA | AK017441.1 | - | 3 | 1066 | ||
AK078818.1 | - | 3 | 880 | |||
BC038308.1 | - | 3 | 717 | |||
BC090967.1 | - | 3 | 1055 | 30652983 | ||
DQ002400.1 | - | 3 | 726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
