siRNA Details
Gene Symbol: Upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026815.2 | NP_081091.1 | 3 | 292 | ||
GenBank mRNA | AF262335.1 | - | 2 | 264 | ||
AK004014.1 | - | 3 | 293 | |||
BC132047.1 | - | 2 | 284 | 40130324 | ||
BC138390.1 | - | 2 | 284 | 8861410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
