siRNA Details
Gene Symbol: Npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023456.2 | NP_075945.1 | 2 | 238 | ||
GenBank mRNA | AF273768.1 | - | 1 | 165 | ||
AF286198.1 | - | 1 | 165 | |||
AF286199.1 | - | 1 | 165 | |||
AK002982.1 | - | 2 | 251 | |||
BC043012.1 | - | 2 | 238 | 5683102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
