siRNA Details
Gene Symbol: Arrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177231.2 | NP_796205.1 | 8, 9 | 916 | ||
NM_178220.3 | NP_835738.1 | 8, 9 | 916 | |||
GenBank mRNA | AK052588.1 | - | 8, 9 | 693 | ||
AK090090.1 | - | 8, 9 | 743 | |||
AK144054.1 | - | 8, 9 | 715 | |||
AK147508.1 | - | 8, 9 | 917 | |||
AK147566.1 | - | 8, 9 | 938 | |||
AK165514.1 | - | 8, 9 | 715 | |||
BC108969.1 | - | 8, 9 | 683 | 40047224 | ||
BC108970.1 | - | 8, 9 | 683 | 40047225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
