siRNA Details
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026678.4 | NP_080954.4 | 8 | 861 | ||
GenBank mRNA | AK002231.1 | - | 7 | 800 | ||
AK010847.1 | - | 8 | 816 | |||
AK172620.1 | - | 8 | 826 | |||
AK182323.1 | - | 3 | 252 | |||
BC052146.1 | - | 8 | 851 | 6588485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
