siRNA Details
Gene Symbol: Pgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025973.3 | NP_080249.2 | 8 | 1038 | ||
GenBank mRNA | AK008886.1 | - | 8 | 1039 | ||
AK008959.1 | - | 8 | 1039 | |||
BC099409.1 | - | 8 | 1029 | 30790663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
