siRNA Details
Gene Symbol: Amy2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042711.2 | NP_001036176.1 | 148 | |||
GenBank mRNA | AK078968.1 | - | 146 | |||
AK133526.1 | - | 149 | ||||
BC094924.1 | - | 136 | 6433956 | |||
BC100579.1 | - | 143 | 6433256 | |||
V00718.1 | - | 149 | ||||
X02576.1 | - | 147 | ||||
X02577.1 | - | 147 | ||||
X02578.1 | - | 147 | ||||
X02579.1 | - | 147 |
Gene Symbol: Amy2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160152.1 | NP_001153624.1 | 1 | 286 |
Gene Symbol: Amy2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160151.1 | NP_001153623.1 | 286 |
Gene Symbol: Amy2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160150.1 | NP_001153622.1 | 286 |
Gene Symbol: Amy2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190403.1 | NP_001177332.1 | 1 | 148 | ||
NM_001190404.1 | NP_001177333.1 | 1 | 148 |

More Information
Other Species Targeted

Gene Ontology Categories:
