siRNA Details
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025900.2 | NP_080176.2 | 7 | 812 | ||
GenBank mRNA | AK007546.1 | - | 4 | 361 | ||
AK012615.1 | - | 7 | 753 | |||
AK012877.1 | - | 7 | 747 | |||
AK033451.1 | - | 7 | 702 | |||
AK141790.1 | - | 1 | 164 | |||
BC055451.1 | - | 7 | 758 | 4164972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
