siRNA Details
Gene Symbol: Syt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138649.1 | NP_619590.1 | 7 | 1342 | ||
GenBank mRNA | AB069667.1 | - | 7 | 1342 | ||
AK135098.1 | - | 5 | 1136 | |||
AK144560.1 | - | 7 | 1519 | |||
AK145293.1 | - | 7 | 1548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
