siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159646.1 | NP_001153118.1 | 2, 3 | 284 | ||
NM_023595.6 | NP_076084.2 | 1, 2 | 379 | |||
GenBank mRNA | AF091101.1 | - | 1, 2 | 236 | ||
AK003076.1 | - | 1, 2 | 168 | |||
AK011407.1 | - | 1, 2 | 170 | |||
AK017216.1 | - | 2, 3 | 274 | |||
AK077244.1 | - | 1, 2 | 412 | |||
AK135438.1 | - | 2, 3 | 293 | |||
AK152459.1 | - | 1, 2 | 175 | |||
AK153285.1 | - | 1, 2 | 160 | |||
BC019979.1 | - | 2, 3 | 266 | 4187559 | ||
BC053693.1 | - | 1, 2 | 166 | 6485182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
