siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159646.1 | NP_001153118.1 | 3 | 310 | ||
NM_023595.6 | NP_076084.2 | 2 | 405 | |||
GenBank mRNA | AF004565.2 | - | 209 | |||
AF091101.1 | - | 2 | 262 | |||
AK003076.1 | - | 2 | 194 | |||
AK011407.1 | - | 2 | 196 | |||
AK017216.1 | - | 3 | 300 | |||
AK077244.1 | - | 2 | 438 | |||
AK135438.1 | - | 3 | 319 | |||
AK152459.1 | - | 2 | 201 | |||
AK153285.1 | - | 2 | 186 | |||
BC019979.1 | - | 3 | 292 | 4187559 | ||
BC053693.1 | - | 2 | 192 | 6485182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
