siRNA Details
Gene Symbol: Hba-a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083955.1 | NP_001077424.1 | 542 | |||
GenBank mRNA | AK010422.1 | - | 543 | |||
AK011030.1 | - | 543 | ||||
AK011073.1 | - | 543 | ||||
AK011121.1 | - | 543 | ||||
AK012348.1 | - | 543 | ||||
AK019138.1 | - | 544 | ||||
CF740193.1 | - | 529 |
Gene Symbol: Hba-a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008218.2 | NP_032244.2 | 3 | 545 | ||
GenBank mRNA | AK010987.1 | - | 3 | 543 | ||
AK011018.1 | - | 3 | 545 | |||
AK011023.1 | - | 3 | 542 | |||
AK011025.1 | - | 3 | 545 | |||
AK011028.1 | - | 3 | 542 | |||
AK011055.1 | - | 3 | 546 | |||
AK011116.1 | - | 1056 | ||||
AK012329.1 | - | 3 | 543 | |||
BC043020.1 | - | 3 | 525 | 5684314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
