siRNA Details
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253712.1 | NP_001240641.1 | 2 | 361 | ||
NM_001253715.1 | NP_001240644.1 | 2 | 347 | |||
NM_001253716.1 | NP_001240645.1 | 2 | 89 | |||
NM_026322.4 | NP_080598.2 | 7 | 760 | |||
GenBank mRNA | AK002356.1 | - | 2 | 236 | ||
AK009822.1 | - | 2 | 238 | |||
AK018338.1 | - | 2 | 83 | |||
AK030278.1 | - | 2 | 239 | |||
AK049714.1 | - | 2 | 362 | |||
BB652761.1 | - | 2 | 359 | |||
BC014738.1 | - | 2 | 306 | 4239349 | ||
BC089311.1 | - | 7 | 760 | 6412625 |

More Information
Other Species Targeted

Gene Ontology Categories:
