siRNA Details
Gene Symbol: Pmch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029971.2 | NP_084247.1 | 1 | 285 | ||
GenBank mRNA | AK020723.1 | - | 1 | 285 | ||
AK138363.1 | - | 1 | 288 | |||
BC048534.1 | - | 1 | 285 | 6533154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
