siRNA Details
Gene Symbol: Tas1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031867.2 | NP_114073.1 | 6 | 2366 | ||
GenBank mRNA | AF301161.1 | - | 6 | 2262 | ||
AF301162.1 | - | 6 | 2262 | |||
AF337040.1 | - | 6 | 2444 | |||
AK153579.1 | - | 6 | 2316 | |||
AY032622.1 | - | 6 | 2262 | |||
BC070448.1 | - | 5 | 4279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
