siRNA Details
Gene Symbol: Ly6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271416.1 | NP_001258345.1 | 451 | |||
NM_001271417.1 | NP_001258346.1 | 436 | ||||
NM_001271418.1 | NP_001258347.1 | 415 | ||||
NM_001271419.1 | NP_001258348.1 | 405 | ||||
NM_001271446.1 | NP_001258375.1 | 431 | ||||
NM_010738.3 | NP_034868.1 | 420 | ||||
GenBank mRNA | AK076200.1 | - | 282 | |||
BC002070.1 | - | 3 | 277 | 3486025 | ||
BG085582.2 | - | 274 | ||||
BY228083.1 | - | 294 | ||||
EU704682.1 | - | 3 | 265 | |||
J03636.1 | - | 284 | ||||
M18184.1 | - | 3 | 251 | |||
X04653.1 | - | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
