siRNA Details
Gene Symbol: Timp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080639.3 | NP_542370.3 | 2 | 284 | ||
GenBank mRNA | AF282730.1 | - | 2 | 215 | ||
AK038820.1 | - | 2 | 283 | |||
AK040814.1 | - | 2 | 283 | |||
AK079347.1 | - | 2 | 285 | |||
AK140367.1 | - | 2 | 284 | |||
BC064046.1 | - | 2 | 264 | 875422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
