siRNA Details
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078167.2 | NP_001071635.1 | 3 | 978 | ||
NM_173374.4 | NP_775550.2 | 3 | 978 | |||
GenBank mRNA | AK018176.1 | - | 3 | 647 | ||
AK053674.1 | - | 3 | 669 | |||
AK078715.1 | - | 3 | 635 | |||
AK150535.1 | - | 3 | 681 | |||
AK169203.1 | - | 3 | 635 | |||
AK169623.1 | - | 3 | 636 | |||
BC046773.1 | - | 3 | 610 | 5708288 | ||
BC058627.1 | - | 3 | 644 | 6401674 | ||
X66091.1 | - | 3 | 591 |

More Information
Other Species Targeted

Gene Ontology Categories:
