siRNA Details
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029546.2 | NP_083822.1 | 3 | 272 | ||
GenBank mRNA | AB041855.1 | - | 3 | 233 | ||
AK088254.1 | - | 3 | 273 | |||
AK088983.1 | - | 3 | 273 | |||
BC112325.1 | - | 3 | 238 | 5355810 | ||
BC117903.1 | - | 3 | 253 | |||
BC128480.1 | - | 3 | 253 | 40097069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
