siRNA Details
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029546.2 | NP_083822.1 | 6, 7 | 668 | ||
GenBank mRNA | AB041855.1 | - | 6, 7 | 629 | ||
AK088254.1 | - | 6, 7 | 669 | |||
AK196838.1 | - | 2, 3 | 234 | |||
BC112325.1 | - | 6, 7 | 634 | 5355810 | ||
BC128480.1 | - | 6, 7 | 649 | 40097069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
