siRNA Details
Gene Symbol: Etfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145615.4 | NP_663590.3 | 2 | 244 | ||
GenBank mRNA | AK028118.1 | - | 2 | 245 | ||
AK032830.1 | - | 2 | 197 | |||
AK133525.1 | - | 2 | 190 | |||
AK167572.1 | - | 2 | 217 | |||
AK168321.1 | - | 2 | 205 | |||
BC003432.1 | - | 2 | 135 | 2646522 | ||
BC096645.1 | - | 2 | 103 | 1245213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
