siRNA Details
Gene Symbol: Mbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033072.2 | NP_149063.2 | 7 | 2171 | ||
GenBank mRNA | AK158797.1 | - | 7 | 2295 | ||
BC059015.1 | - | 8 | 1883 | |||
BC096027.1 | - | 4 | 1913 |

More Information
Other Species Targeted

Gene Ontology Categories:
