siRNA Details
Gene Symbol: Erc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053204.2 | NP_444434.2 | 3 | 1101 | ||
NM_178085.3 | NP_835186.1 | 2 | 372 | |||
GenBank mRNA | AF340028.1 | - | 3 | 1085 | ||
AF340029.1 | - | 3 | 1085 | |||
AK035989.1 | - | 2 | 915 | |||
AK048990.1 | - | 2 | 325 | |||
AK053824.1 | - | 2 | 912 | |||
AK134383.1 | - | 1 | 1940 | |||
AY316692.1 | - | 1 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
