siRNA Details
Gene Symbol: Dxo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163770.1 | NP_001157242.1 | 7 | 1339 | ||
NM_033613.2 | NP_291091.2 | 7 | 1364 | |||
GenBank mRNA | BC004713.1 | - | 7 | 1300 | 3499698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
