siRNA Details
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053208.4 | NP_444438.2 | 3 | 1325 | ||
GenBank mRNA | AF340231.1 | - | 2 | 1067 | ||
AF453879.1 | - | 3 | 1247 | |||
AJ310547.1 | - | 3 | 1296 | |||
AK075582.1 | - | 3 | 1277 | |||
AK152056.1 | - | 3 | 1286 | |||
AK152223.1 | - | 3 | 1283 | |||
AK159206.1 | - | 3 | 1278 | |||
AK166661.1 | - | 4 | 1191 | |||
AK167477.1 | - | 3 | 1278 | |||
BC023299.1 | - | 3 | 1259 | 5135579 | ||
BC051436.1 | - | 2 | 1043 | 6312638 | ||
BC086764.1 | - | 3 | 1264 | 5685311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
