Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Naphthyl)-2-thiourea 98.0+%, TCI America™
SDP

Catalog No. p-7133599 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 86-88-4
Molecular Formula C11H10N2S
Molecular Weight (g/mol) 202.275
MDL Number MFCD00041824
InChI Key PIVQQUNOTICCSA-UHFFFAOYSA-N
Synonym 1-1-naphthyl-2-thiourea, antu, 1-naphthylthiourea, dirax, anturat, rattrack, smeesana, alrato, bantu, kripid
PubChem CID 736366
ChEBI CHEBI:82256
IUPAC Name naphthalen-1-ylthiourea
SMILES C1=CC=C2C(=C1)C=CC=C2NC(=S)N

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 5 g
UN Number 1651
Formula Weight 202.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(1-Naphthyl)-2-thiourea
RTECSNumber YT9275000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.