Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Decanediol 97.0+%, TCI America™
SDP

Catalog No. p-7125983 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 112-47-0
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.284
MDL Number MFCD00004749
InChI Key FOTKYAAJKYLFFN-UHFFFAOYSA-N
Synonym 1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
PubChem CID 37153
IUPAC Name decane-1,10-diol
SMILES C(CCCCCO)CCCCO

Specifications

Melting Point 73°C
Color White
Boiling Point 192°C
Quantity 25 g
Formula Weight 174.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Decanediol
EINECSNumber (2)-0240
RTECSNumber HD8433713
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.