Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,13-Tetradecadiene, 90+%, Thermo Scientific Chemicals

Catalog No. p-7035752
Click to view available options
Quantity:
1 g
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21964-49-8
Molecular Formula C14H26
Molecular Weight (g/mol) 194.362
MDL Number MFCD00014941
InChI Key XMRSTLBCBDIKFI-UHFFFAOYSA-N
Synonym 1,13-tetradecadiene, tetradecadiene-1,13, acmc-209fpz
PubChem CID 30875
IUPAC Name tetradeca-1,13-diene
SMILES C=CCCCCCCCCCCC=C

Specifications

Density 0.849
Boiling Point 131°C (17 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.443
Linear Formula CH2=CH(CH2)10CH=CH2
Quantity 1 g
Beilstein 1741459
Formula Weight 194.36
Percent Purity ≥90%
Chemical Name or Material 1,13-Tetradecadiene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-687-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.