Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine Copper(II) (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. p-7130920 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 14916-87-1
Molecular Formula C32H2CuF16N8
Molecular Weight (g/mol) 865.944
MDL Number MFCD00274642
InChI Key NKBVANSQKHVZHF-UHFFFAOYSA-N
Synonym Copper(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine, F16CuPc
PubChem CID 87077853
SMILES C12=C(C(=C(C(=C1F)F)F)F)C3=NC4=NC(=NC5=NC(=NC6=C7C(=C(N6)N=C2N3)C(=C(C(=C7F)F)F)F)C8=C5C(=C(C(=C8F)F)F)F)C9=C4C(=C(C(=C9F)F)F)F.[Cu]

Specifications

Color Purple-Red
Quantity 100 mg
Formula Weight 863.93
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine Copper(II) (purified by subli
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.