Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,5-Trimethyl-1H-pyrrole-3-carboxylic acid, Technical Grade, Thermo Scientific™

Catalog No. p-4509976
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 175276-50-3
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00203866
InChI Key JTEBLTWGSAXWEE-UHFFFAOYSA-N
Synonym 1,2,5-trimethyl-1h-pyrrole-3-carboxylic acid, maybridge1_002499, 3-carboxy-1,2,5-trimethyl-1h-pyrrole, 1,2,5-trimethyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid,1,2,5-trimethyl, 1h-pyrrole-3-carboxylicacid, 1,2,5-trimethyl, 1h-pyrrole-3-carboxylic acid, 1,2,5-trimethyl
PubChem CID 736491
IUPAC Name 1,2,5-trimethylpyrrole-3-carboxylic acid
SMILES CN1C(C)=CC(C(O)=O)=C1C

Specifications

Quantity 250 mg
Formula Weight 153.18
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1, 2, 5-Trimethyl-1H-pyrrole-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.