Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic Acid 90.0+%, TCI America™
SDP

Catalog No. p-7123231 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 85233-19-8
Molecular Formula C22H24N2O10
Molecular Weight (g/mol) 476.44
MDL Number MFCD00036255
InChI Key FTEDXVNDVHYDQW-UHFFFAOYSA-N
Synonym bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid
PubChem CID 104751
ChEBI CHEBI:60888
IUPAC Name 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid
SMILES OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O

Specifications

Melting Point 159°C
Color White
Quantity 100 mg
Formula Weight 476.44
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic Acid
RTECSNumber MC0423000
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.