Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(dimethylphosphino)ethane 97.0+%, TCI America™
SDP

Catalog No. p-7118899 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 23936-60-9
Molecular Formula C6H16P2
Molecular Weight (g/mol) 150.14
MDL Number MFCD00008511
InChI Key ZKWQSBFSGZJNFP-UHFFFAOYSA-N
Synonym 1,2-bis dimethylphosphino ethane, ethylenebis dimethylphosphine, phosphine, 1,2-ethanediylbis dimethyl, 2-dimethylphosphanylethyl dimethyl phosphane, dimethyl-2-dimethylphosphino ethylphosphine, 2-dimethylphosphanyl ethyl dimethylphosphane, pubchem6482, acmc-1cjp0, ghl.pd_mitscher_leg0.173, zkwqsbfsgzjnfp-uhfffaoysa
PubChem CID 141059
IUPAC Name [2-(dimethylphosphanyl)ethyl]dimethylphosphane
SMILES CP(C)CCP(C)C

Specifications

Color Colorless
Boiling Point 180°C
Quantity 1 g
UN Number 1993
Formula Weight 150.14
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(dimethylphosphino)ethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.