Learn More
1,2-Bis(diphenylphosphino)ethane, 97+%, Thermo Scientific Chemicals
Description
1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,2-Bis(diphenylphosphino)ethane is a bidendate ligand used in coordination chemistry to prepare metal complexes. It is involved in the metal-catalyzed allylic alkylation and decarboxylation of allylic esters. It is also used in 1,3-diene synthesis. Further, it is used in cycloaddition reactions and carbonylation reactions. In addition to this, it is used to prepare bis(dicyclohexylphosphino)ethane by hydrogenation.
Solubility
Soluble in acetone, methanol and alcoholic hydrochloric acid.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 138°C to 143°C |
Linear Formula | (C6H5)2PCH2CH2P(C6H5)2 |
Quantity | 5 g |
Beilstein | 761261 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in acetone,methanol and alcoholic hydrochloric acid. |
Formula Weight | 398.42 |
Percent Purity | ≥97% |
Chemical Name or Material | 1,2-Bis (diphenylphosphino)ethane |
Safety and Handling
Hazard Category | H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 216-769-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.