Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Cyclododecanediol (cis- and trans- mixture) 87.0+%, TCI America™
SDP

Catalog No. C14971G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 15199-41-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD01321150
InChI Key HAMFVYJFVXTJCJ-UHFFFAOYSA-N
PubChem CID 85817
IUPAC Name cyclododecane-1,2-diol
SMILES C1CCCCCC(C(CCCC1)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 200.32
Percent Purity ≥87.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Cyclododecanediol (cis- and trans- mixture)
EINECSNumber (3)-2291
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.