Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dihydronaphthalene 98.0+%, TCI America™
SDP

Catalog No. p-7126302 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 447-53-0
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00001672
InChI Key KEIFWROAQVVDBN-UHFFFAOYSA-N
Synonym dialin, 1,2-dialin, diolin, delta1-dialin, dihydronaphthalene, naphthalene, 1,2-dihydro, dialin van, delta 1-dialin, dihydronapthalene, naphthalene, dihydro
PubChem CID 9938
ChEBI CHEBI:38142
IUPAC Name 1,2-dihydronaphthalene
SMILES C1CC2=CC=CC=C2C=C1

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 207°C
Quantity 1 g
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dihydronaphthalene
EINECSNumber (4)-1177
RTECSNumber QJ4375000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.