Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Nitrophenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. p-7134034 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 59084-06-9
Molecular Formula C10H14N3O2
Molecular Weight (g/mol) 208.24
MDL Number MFCD00040728
InChI Key YJRCDSXLKPERNV-UHFFFAOYSA-O
Synonym 1-2-nitrophenyl piperazine, 1-2-nitro-phenyl-piperazine, 2-nitrophenyl piperazine, 1-2-nitrophenyl piperazin, nitrophenylpiperazine, acmc-20ahsu, pubchem14574, 4-nitrophenyl piperazine
PubChem CID 100949
IUPAC Name 4-(2-nitrophenyl)piperazin-1-ium
SMILES [O-][N+](=O)C1=CC=CC=C1N1CC[NH2+]CC1

Specifications

Boiling Point 126°C
Quantity 5 g
Formula Weight 207.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Nitrophenyl)piperazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.