Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ 1,2-Propanediol
Shop All AquaPhoenix Scientific Products

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Solution
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
Color | Colorless |
Density | 1.0361g/cmsuperscript_3; |
Assay Percent Range | 96% w/v |
Packaging | Solution |
Linear Formula | CH3CHOHCH2OH |
Quantity | 100 mL |
Formula Weight | 76.1 |
Chemical Name or Material | 1, 2-Propanediol |
Safety and Handling

ShelfLife | 2 years |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction