Learn More
Ace Glass, Inc. Flask, freeze drying, 1.2L, 249mm height, 53mm ID neck, 100mm body OD, includes snap-on rubber cap and 1/2in OD PTFE plain manifold connection adapter
Supplier:  Ace Glass, Inc. 703527
Encompass_Preferred
Catalog No. 50-979-82
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.