Learn More
Trajan Scientific and Medical™ 1.2mm ID Direct Straight-through Liners
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Supplier: Trajan Scientific and Medical™ 092016
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent and Thermo Scientific instruments
Specifications
GC Inlet Liner, Direct, Straight-Through Liner | |
5/Pk. | |
6.3 mm |
78.5 mm | |
1.2 mm | |
Agilent and Thermo Scientific Instruments |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.