Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzodioxol-4-ylmethanol, 97%, Thermo Scientific™

Catalog No. CC01709DA
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

769-30-2

Molecular Formula

C8H8O3

Molecular Weight (g/mol)

152.149

MDL Number

MFCD02681980

InChI Key

XVCMMPXFVAHHQN-UHFFFAOYSA-N

Synonym

benzo d 1,3 dioxol-4-ylmethanol, 1,3-benzodioxole-4-methanol, 2h-1,3-benzodioxol-4-ylmethanol, benzo 1,3 dioxol-4-yl-methanol, 1,3-benzodioxol-4-yl-methanol, 1,3-benzodioxol-4-yl methanol, 2,3-methylenedioxy benzyl alcohol, 4-hydroxymethyl-1,3-benzodioxole, benzo d 1,3 dioxol-7-yl methanol, 2h-benzo d 1,3-dioxolen-4-ylmethan-1-ol

PubChem CID

2776187

IUPAC Name

1,3-benzodioxol-4-ylmethanol

SMILES

C1OC2=CC=CC(=C2O1)CO

Specifications

CAS Min %

96.5

CAS Max %

100.0

Assay Percent Range

97%

Packaging

Amber Glass Bottle

Quantity

1 g

Formula Weight

152.15

Percent Purity

97%

Chemical Name or Material

1, 3-Benzodioxol-4-ylmethanol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.