Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzodioxol-4-ylmethanol, 97%, Thermo Scientific™

Catalog No. p-2694628
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 769-30-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD02681980
InChI Key XVCMMPXFVAHHQN-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxol-4-ylmethanol, 1,3-benzodioxole-4-methanol, 2h-1,3-benzodioxol-4-ylmethanol, benzo 1,3 dioxol-4-yl-methanol, 1,3-benzodioxol-4-yl-methanol, 1,3-benzodioxol-4-yl methanol, 2,3-methylenedioxy benzyl alcohol, 4-hydroxymethyl-1,3-benzodioxole, benzo d 1,3 dioxol-7-yl methanol, 2h-benzo d 1,3-dioxolen-4-ylmethan-1-ol
PubChem CID 2776187
IUPAC Name 1,3-benzodioxol-4-ylmethanol
SMILES C1OC2=CC=CC(=C2O1)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material 1, 3-Benzodioxol-4-ylmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.