Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzodioxole-5-sulfonyl chloride, 95%, Thermo Scientific™

Catalog No. CC01603DE
Click to view available options
Quantity:
Packaging:

Chemical Identifiers

CAS

115010-10-1

Molecular Formula

C7H5ClO4S

Molecular Weight (g/mol)

220.623

InChI Key

ICUBASIDCXDQAW-UHFFFAOYSA-N

Synonym

benzo d 1,3 dioxole-5-sulfonyl chloride, benzo 1,3 dioxole-5-sulfonyl chloride, 2h-1,3-benzodioxole-5-sulfonyl chloride, 1,3-benzodioxole-5-sulfonylchloride, 1,3-benzodioxole-5-sulphonyl chloride, acmc-20a4hu, 5-chlorosulphonyl-1,3-benzodioxole, benzo 1,3 dioxol-5-sulfonyl chloride, benzo 1,3 dioxol-5-sulphonyl chloride, benzo 1,3-dioxol-5-sulphonyl chloride

PubChem CID

4913401

IUPAC Name

1,3-benzodioxole-5-sulfonyl chloride

SMILES

C1OC2=C(O1)C=C(C=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range

95%

Packaging

Amber Glass Bottle

Quantity

5 g

Formula Weight

220.63

Percent Purity

95%

Chemical Name or Material

1, 3-Benzodioxole-5-sulfonyl chloride

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.