Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzodioxole-5-sulfonyl chloride, 95%, Thermo Scientific™

Catalog No. p-4509830
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 115010-10-1
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.623
InChI Key ICUBASIDCXDQAW-UHFFFAOYSA-N
Synonym benzo d 1,3 dioxole-5-sulfonyl chloride, benzo 1,3 dioxole-5-sulfonyl chloride, 2h-1,3-benzodioxole-5-sulfonyl chloride, 1,3-benzodioxole-5-sulfonylchloride, 1,3-benzodioxole-5-sulphonyl chloride, acmc-20a4hu, 5-chlorosulphonyl-1,3-benzodioxole, benzo 1,3 dioxol-5-sulfonyl chloride, benzo 1,3 dioxol-5-sulphonyl chloride, benzo 1,3-dioxol-5-sulphonyl chloride
PubChem CID 4913401
IUPAC Name 1,3-benzodioxole-5-sulfonyl chloride
SMILES C1OC2=C(O1)C=C(C=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 220.63
Percent Purity 95%
Chemical Name or Material 1, 3-Benzodioxole-5-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.