Learn More
AdipoGen 1 3-Cyclohexadiene Stabil BHT
Chemical. CAS 592-57-4. Formula C6H8. MW 80.1. 1,3-Cyclohexadiene is an important building block and synthesis for organic synthesis. It is used by example as a hydrogen donor in transfer hydrogenation and in the conversion to benzene. Contains 3ppm BHT as a stabilizer.