Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dicyclohexylthiourea 98.0+%, TCI America™
SDP

Catalog No. p-7126214 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1212-29-9
Molecular Formula C13H24N2S
Molecular Weight (g/mol) 240.409
MDL Number MFCD00021316
InChI Key KAJICSGLHKRDLN-UHFFFAOYSA-N
Synonym n,n'-dicyclohexylthiourea, thiourea, n,n'-dicyclohexyl, 1,3-dicyclohexyl-2-thiourea, sym-dicyclohexylthiourea, dicyclohexyl thiourea, sym-dicyclohexythiourea, 1,3-bis cyclohexyl thiourea, n,n'-dicyclohexylthiocarbamide, n,n'-dicyclohexyl-2-thiourea, dicyclohexylthiourea
PubChem CID 727200
IUPAC Name 1,3-dicyclohexylthiourea
SMILES C1CCC(CC1)NC(=S)NC2CCCCC2

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 5 g
Formula Weight 240.41
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dicyclohexylthiourea
EINECSNumber (3)-2497
RTECSNumber YS9300000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.